Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TTN antisense RNA 1 | TTN-AS1 | -103.2kb | Sense | Chr. 2 - 178522827 - 178620217 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101927055 | LOC101927055 | 49.9kb | Sense | Chr. 2 - 178776926 - 178779963 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map