Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | runt related transcription factor 3 | RUNX3 |
AML2, CBFA3, PEBP2aC |
-3.3kb | Sense | Chr. 1 - 24899511 - 24965158 on NCBI Build GRCh38 |
| Downstream | microRNA 4425 | MIR4425 | 52.6kb | Anti-sense | Chr. 1 - 25023503 - 25023586 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map