Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Ewing tumor associated antigen 1 | ETAA1 | ETAA16 | -298.7kb | Sense | Chr. 2 - 67397310 - 67410411 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927701 | LOC101927701 | 84.4kb | Anti-sense | Chr. 2 - 67796054 - 67825562 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map