Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chaperonin containing TCP1 subunit 6 pseudogene 3 | CCT6P3 | CCT6-3P | -3.9kb | Anti-sense | Chr. 7 - 65038354 - 65074713 on NCBI Build GRCh38 |
| Downstream | integrator complex subunit 4 pseudogene 1 | INTS4P1 | INTS4L1 | 59.9kb | Anti-sense | Chr. 7 - 65141037 - 65197161 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map