Assay Details
| Gene Symbol: LOC101927441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC045184.1 | - | 1 - 2 | 151 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | forkhead associated phosphopeptide binding domain 1 | FHAD1 | -8.8kb | Anti-sense | Chr. 1 - 15236559 - 15400283 on NCBI Build GRCh38 | |
| Downstream | EF-hand domain family member D2 | EFHD2 | SWS1 | 461.0 | Anti-sense | Chr. 1 - 15409895 - 15430343 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012998.hg.1 | ENST00000427824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012998.hg.1 | ENST00000427824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012998.hg.1 | ENST00000427824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012998.hg.1 | ENST00000427824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012998.hg.1 | ENST00000427824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012998.hg.1 | ENST00000427824.1 | Ensembl |

LOC101927441
Genomic Map