Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105371910 | LOC105371910 | -2.4kb | Anti-sense | Chr. 17 - 78248316 - 78252491 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100996291 | LOC100996291 | 3.5kb | Sense | Chr. 17 - 78261349 - 78278492 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map