Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SNAP25 antisense RNA 1 | SNAP25-AS1 | -68.5kb | Anti-sense | Chr. 20 - 10023812 - 10219506 on NCBI Build GRCh38 | |
| Downstream | McKusick-Kaufman syndrome | MKKS |
BBS6, HMCS, KMS, MKS |
114.1kb | Anti-sense | Chr. 20 - 10404780 - 10434239 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map