Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glycophorin A (MNS blood group) | GYPA |
CD235a, GPA, GPErik, GPSAT, HGpMiV, HGpMiXI, HGpSta(C), MN, MNS, PAS-2 |
-365.1kb | Anti-sense | Chr. 4 - 144109303 - 144140854 on NCBI Build GRCh38 |
| Downstream | HHIP antisense RNA 1 | HHIP-AS1 | 133.9kb | Anti-sense | Chr. 4 - 144642916 - 144646019 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map