Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mediator complex subunit 4 | MED4 |
ARC36, DRIP36, HSPC126, TRAP36, VDRIP |
-161.9kb | Anti-sense | Chr. 13 - 48075728 - 48095141 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 441 | LINC00441 | ncRNA-RB1 | 35.1kb | Anti-sense | Chr. 13 - 48296513 - 48303661 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map