Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
NSE3 homolog, SMC5-SMC6 complex component |
NSMCE3 |
HCA4, MAGEG1, MAGEL3, NDNL2, NSE3 |
-103.8kb | Sense | Chr. 15 - 29268149 - 29269816 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100130111 | LOC100130111 | 298.0kb | Anti-sense | Chr. 15 - 29674935 - 29679163 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map