Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MIR31 host gene | MIR31HG | -299.2kb | Sense | Chr. 9 - 21454268 - 21559698 on NCBI Build GRCh38 | |
| Downstream | endogenous retrovirus group FRD member 3 | ERVFRD-3 | 67.5kb | Anti-sense | Chr. 9 - 21929257 - 21931273 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map