| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000229465.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000229465.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000229465.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000229465.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000229465.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000229465.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000229465.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000556569.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000556569.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000556569.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000556569.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000556569.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000556569.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000556569.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000556717.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000556717.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000556717.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000556717.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
ENST00000556717.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
NR_039985.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
NR_039985.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
NR_039985.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
NR_039985.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
NR_039985.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
NR_039985.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
NR_039985.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010618.hg.1 |
NR_039985.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
ENST00000508827.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
ENST00000508827.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
ENST00000508827.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
ENST00000508827.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
ENST00000508827.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
ENST00000508827.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
ENST00000508827.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
ENST00000508827.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
ENST00000553288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
ENST00000553288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
ENST00000553288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
ENST00000553288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
ENST00000553288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
ENST00000553288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
ENST00000553288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
ENST00000553288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
NR_110416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
NR_110416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
NR_110416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
NR_110416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
NR_110416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
NR_110416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
NR_110416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
NR_110416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
NR_110417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
NR_110417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
NR_110417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009364.hg.1 |
NR_110417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000229465.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000229465.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000229465.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000229465.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000229465.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000229465.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000229465.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000556569.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000556569.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000556569.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000556569.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000556569.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000556569.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000556569.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000556717.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000556717.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000556717.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000556717.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
ENST00000556717.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
NR_039985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
NR_039985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
NR_039985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
NR_039985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
NR_039985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
NR_039985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
NR_039985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010618.hg.1 |
NR_039985.1 |
Refseq |