Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ret finger protein like 2 | RFPL2 | RNF79 | -119.0kb | Sense | Chr. 22 - 32190435 - 32204844 on NCBI Build GRCh38 |
| Downstream | ret finger protein like 3 | RFPL3 | 25.3kb | Anti-sense | Chr. 22 - 32354885 - 32361161 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map