Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | LARGE xylosyl- and glucuronyltransferase 1 | LARGE1 |
LARGE, MDC1D, MDDGA6, MDDGB6 |
-167.4kb | Anti-sense | Chr. 22 - 33162237 - 33922841 on NCBI Build GRCh38 |
| Downstream | intestine specific homeobox | ISX |
Pix-1, RAXLX |
970.9kb | Sense | Chr. 22 - 35066136 - 35087387 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map