Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
apoptosis inducing factor, mitochondria associated 2 |
AIFM2 |
AMID, PRG3 |
Anti-sense | Chr. 10 - 70112267 - 70132934 on NCBI Build GRCh38 | |
| Downstream | trypsin domain containing 1 | TYSND1 | NET41 | 16.7kb | Anti-sense | Chr. 10 - 70137981 - 70146740 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map