Assay Details
| Gene Symbol: MAP3K13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242314.1 | NP_001229243.1 | 15 | 7823 | 104 | |
| NM_001242317.1 | NP_001229246.1 | 13 | 7087 | 104 | ||
| NM_004721.4 | NP_004712.1 | 14 | 7938 | 104 | ||
| XM_017007456.1 | XP_016862945.1 | 15 | 7834 | 104 | ||
| GenBank mRNA | AK123148.1 | - | 1 | 49 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 41A | TMEM41A | 2900010K02Rik | Anti-sense | Chr. 3 - 185482671 - 185499057 on NCBI Build GRCh38 | |
| Downstream | lipase H | LIPH |
AH, ARWH2, HYPT7, LAH2, LPDLR, PLA1B, mPA-PLA1 |
17.8kb | Anti-sense | Chr. 3 - 185506262 - 185552661 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MAP3K13
Genomic Map