Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MIR3663 host gene | MIR3663HG | -6.0kb | Anti-sense | Chr. 10 - 117153521 - 117169055 on NCBI Build GRCh38 | |
| Downstream | potassium two pore domain channel subfamily K member 18 | KCNK18 |
K2p18.1, MGR13, TRESK, TRESK-2, TRESK2, TRIK |
20.1kb | Sense | Chr. 10 - 117197489 - 117210299 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map