Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4717 | MIR4717 | -60.2kb | Anti-sense | Chr. 16 - 2274620 - 2274691 on NCBI Build GRCh38 | |
| Downstream |
ATP binding cassette subfamily A member 17, pseudogene |
ABCA17P | ABCA17 | 3.9kb | Anti-sense | Chr. 16 - 2340922 - 2426699 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map