| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009932.hg.1 |
ENST00000563601.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009932.hg.1 |
NR_110370.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009932.hg.1 |
NR_110370.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009932.hg.1 |
ENST00000563601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009932.hg.1 |
NR_110370.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009932.hg.1 |
NR_110370.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000167912.hg.1 |
NR_110370.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000167912.hg.1 |
NR_110370.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000167912.hg.1 |
TC1000009932.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000167911.hg.1 |
ENST00000563601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000167911.hg.1 |
NR_110370.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000167911.hg.1 |
ENST00000563601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000167911.hg.1 |
NR_110370.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000167911.hg.1 |
TC1000009932.hg.1 |
Clariom D TC |