Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2 |
WFIKKN2 |
GASP-1, WFDC20B, WFIKKNRP, hGASP-1 |
-25.0kb | Sense | Chr. 17 - 50834650 - 50842353 on NCBI Build GRCh38 |
| Downstream | sperm associated antigen 9 | SPAG9 |
CT89, HLC-6, HLC4, HLC6, JIP-4, JIP4, JLP, PHET, PIG6 |
92.8kb | Anti-sense | Chr. 17 - 50962174 - 51120865 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map