Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uridine monophosphate synthetase | UMPS | OPRT | -41.8kb | Anti-sense | Chr. 3 - 124730366 - 124749273 on NCBI Build GRCh38 |
| Downstream |
mucin 13, cell surface associated |
MUC13 |
DRCC1, MUC-13 |
112.3kb | Sense | Chr. 3 - 124905442 - 124934751 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map