Assay Details
| Gene Symbol: TTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005271277.3 | XP_005271334.1 | 23 | 9179 | 82 | |
| XM_011542303.2 | XP_011540605.1 | 23 | 9203 | 82 | ||
| XM_017002549.1 | XP_016858038.1 | 24 | 9266 | 82 | ||
| XM_017002550.1 | XP_016858039.1 | 25 | 8635 | 82 | ||
| XM_017002551.1 | XP_016858040.1 | 25 | 8562 | 82 | ||
| XM_017002552.1 | XP_016858041.1 | 24 | 9263 | 82 | ||
| XM_017002553.1 | XP_016858042.1 | 24 | 9245 | 82 | ||
| XM_017002554.1 | XP_016858043.1 | 24 | 9242 | 82 | ||
| XM_017002555.1 | XP_016858044.1 | 24 | 8551 | 82 | ||
| GenBank mRNA | AK123765.1 | - | 1 | 1847 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Downstream | tripartite motif containing 45 | TRIM45 | RNF99 | 3.6kb | Anti-sense | Chr. 1 - 117107711 - 117121789 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TTF2
Genomic Map