Assay Details
| Gene Symbol: KCTD13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017023106.1 | XP_016878595.1 | 3 | 2131 | 83 | |
| GenBank mRNA | AK123825.1 | - | 1 | 1447 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | aspartate beta-hydroxylase domain containing 1 | ASPHD1 | Anti-sense | Chr. 16 - 29900392 - 29919860 on NCBI Build GRCh38 | ||
| Downstream | transmembrane protein 219 | TMEM219 | IGFBP-3R | 40.1kb | Anti-sense | Chr. 16 - 29962030 - 29973052 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

KCTD13
Genomic Map