Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
FTX transcript, XIST regulator (non-protein coding) |
FTX |
LINC00182, MIR374AHG, NCRNA00182 |
Sense | Chr. X - 74028136 - 74293574 on NCBI Build GRCh38 | |
| Downstream | microRNA 421 | MIR421 |
MIRN421, hsa-mir-421 |
15.4kb | Sense | Chr. X - 74218377 - 74218461 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map