Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ArfGAP with coiled-coil, ankyrin repeat and PH domains 2 |
ACAP2 |
CENTB2, CNT-B2 |
-78.3kb | Sense | Chr. 3 - 195274736 - 195443176 on NCBI Build GRCh38 |
| Downstream | apolipoprotein D | APOD | 45.0kb | Sense | Chr. 3 - 195568702 - 195584205 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map