Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protein phosphatase 2 regulatory subunit B''alpha | PPP2R3A |
PPP2R3, PR130, PR72 |
Sense | Chr. 3 - 135965673 - 136147910 on NCBI Build GRCh38 | |
| Downstream | male-specific lethal 2 homolog (Drosophila) | MSL2 |
MSL-2, MSL2L1, RNF184 |
4.3kb | Anti-sense | Chr. 3 - 136148918 - 136197300 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map