Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leukocyte immunoglobulin like receptor A4 | LILRA4 |
CD85g, ILT7 |
-12.2kb | Sense | Chr. 19 - 54333185 - 54339169 on NCBI Build GRCh38 |
| Downstream | leukocyte associated immunoglobulin like receptor 1 | LAIR1 |
CD305, LAIR-1 |
339.0 | Sense | Chr. 19 - 54353624 - 54370556 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map