Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 136 member A | FAM136A | -99.9kb | Sense | Chr. 2 - 70295976 - 70302088 on NCBI Build GRCh38 | |
| Downstream | transforming growth factor alpha | TGFA | TFGA | 43.5kb | Sense | Chr. 2 - 70447280 - 70554015 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map