| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053886.hg.1 |
ENST00000421530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053886.hg.1 |
NR_038072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053886.hg.1 |
TC01003459.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016240.hg.1 |
ENST00000421530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016240.hg.1 |
ENST00000421530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016240.hg.1 |
ENST00000421530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016240.hg.1 |
ENST00000421530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016240.hg.1 |
NR_038072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016240.hg.1 |
NR_038072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016240.hg.1 |
NR_038072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016240.hg.1 |
NR_038072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053887.hg.1 |
ENST00000421530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053887.hg.1 |
NR_038072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053887.hg.1 |
TC01003459.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100223625.hg.1 |
ENST00000421530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100223625.hg.1 |
NR_038072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100223625.hg.1 |
ENST00000421530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100223625.hg.1 |
NR_038072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100223625.hg.1 |
TC0100016240.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100223626.hg.1 |
ENST00000421530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100223626.hg.1 |
NR_038072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100223626.hg.1 |
ENST00000421530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100223626.hg.1 |
NR_038072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100223626.hg.1 |
TC0100016240.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100016240.hg.1 |
ENST00000421530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016240.hg.1 |
ENST00000421530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016240.hg.1 |
ENST00000421530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016240.hg.1 |
ENST00000421530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016240.hg.1 |
NR_038072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016240.hg.1 |
NR_038072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016240.hg.1 |
NR_038072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016240.hg.1 |
NR_038072.1 |
Refseq |