Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeats and immunoglobulin like domains 3 | LRIG3 | LIG3 | -692.1kb | Anti-sense | Chr. 12 - 58869351 - 58920540 on NCBI Build GRCh38 |
| Downstream |
family with sequence similarity 19 member A2, C-C motif chemokine like |
FAM19A2 |
TAFA-2, TAFA2 |
2.1mb | Anti-sense | Chr. 12 - 61708248 - 62192839 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map