Assay Details
| Gene Symbol: ZNF444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011527068.1 | XP_011525370.1 | 1 | 149 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 787 | ZNF787 | TIP20 | -20.8kb | Anti-sense | Chr. 19 - 56087363 - 56121373 on NCBI Build GRCh38 |
| Downstream | galanin like peptide | GALP | 31.7kb | Sense | Chr. 19 - 56176020 - 56185775 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the XM_011527068.1 transcripts.
This assay is in the 5'UTR region of the XM_011527068.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000586123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000586123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000586123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000586123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000588203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000588203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000588203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000588358.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000588358.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000588358.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000588358.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000591564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000591564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000591564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000591564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000586123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000586123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000586123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000586123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000588203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000588203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000588203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000588358.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000588358.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000588358.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000588358.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000591564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000591564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000591564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000591564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |

ZNF444
Genomic Map