Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | xyloside xylosyltransferase 1 | XXYLT1 | C3orf21 | -8.5kb | Sense | Chr. 3 - 195068276 - 195271166 on NCBI Build GRCh38 |
| Downstream | protein phosphatase 1 regulatory inhibitor subunit 2 | PPP1R2 |
IPP-2, IPP2 |
230.5kb | Sense | Chr. 3 - 195514422 - 195543401 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map