Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | intraflagellar transport 122 | IFT122 |
CED, CED1, SPG, WDR10, WDR10p, WDR140 |
Anti-sense | Chr. 3 - 129440036 - 129520507 on NCBI Build GRCh38 | |
| Downstream | rhodopsin | RHO |
CSNBAD1, OPN2, RP4 |
21.1kb | Anti-sense | Chr. 3 - 129528639 - 129535344 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map