Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein L13a pseudogene 6 | RPL13AP6 |
RPL13A_7_1096, bA348N5.5 |
-6.8kb | Anti-sense | Chr. 10 - 110936603 - 110937255 on NCBI Build GRCh38 |
| Downstream | microRNA 548e | MIR548E |
MIRN548E, hsa-mir-548e |
41.2kb | Sense | Chr. 10 - 110988926 - 110989013 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map