Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102723521 | LOC102723521 | -8.0kb | Anti-sense | Chr. 6 - 34273202 - 34282446 on NCBI Build GRCh38 | |
| Downstream | ribosomal protein S10 | RPS10 |
DBA9, S10 |
124.9kb | Sense | Chr. 6 - 34417454 - 34426125 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map