Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TCEB3 antisense RNA 1 | TCEB3-AS1 | Sense | Chr. 1 - 23760382 - 23778297 on NCBI Build GRCh38 | ||
| Downstream | PITH domain containing 1 | PITHD1 |
C1orf128, HT014, TXNL1CL |
15.5kb | Anti-sense | Chr. 1 - 23778386 - 23788232 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map