| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000436594.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000436594.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000436594.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000436594.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000608972.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000608972.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000608972.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000608972.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609495.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609495.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609495.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609495.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609495.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
ENST00000609495.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002817.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002817.hg.1 |
TC07000180.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002816.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002816.hg.1 |
TC07000180.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700148857.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700148857.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700148857.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700148857.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700148857.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700148857.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700148857.hg.1 |
TC0700007038.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700148858.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700148858.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700148858.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700148858.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700148858.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700148858.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700148858.hg.1 |
TC0700007038.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000436594.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000436594.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000436594.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000436594.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000608972.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000608972.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000608972.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000608972.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609495.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609495.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609495.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609495.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609495.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
ENST00000609495.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007038.hg.1 |
NR_038965.1 |
Refseq |