Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929123 | LOC101929123 | -302.3kb | Sense | Chr. 4 - 17171757 - 17186057 on NCBI Build GRCh38 | |
| Downstream | clarin 2 | CLRN2 | 24.6kb | Anti-sense | Chr. 4 - 17515165 - 17527104 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map