Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DiGeorge syndrome critical region gene 5 (non-protein coding) | DGCR5 |
LINC00037, NCRNA00037 |
Sense | Chr. 22 - 18970498 - 19031242 on NCBI Build GRCh38 | |
| Downstream | DiGeorge syndrome critical region gene 9 (non-protein coding) | DGCR9 | DGS-A | 241.0 | Sense | Chr. 22 - 19017834 - 19020248 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map