Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
proteasome 26S subunit, non-ATPase 7 |
PSMD7 |
MOV34, P40, Rpn8, S12 |
-7.7kb | Anti-sense | Chr. 16 - 74296775 - 74306288 on NCBI Build GRCh38 |
| Downstream | pyruvate dehydrogenase phosphatase regulatory subunit pseudogene | LOC283922 | 15.9kb | Sense | Chr. 16 - 74332406 - 74368255 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map