Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sperm acrosome associated 7 | SPACA7 | C13orf28 | -168.1kb | Anti-sense | Chr. 13 - 112376300 - 112434695 on NCBI Build GRCh38 |
| Downstream | ATP11A upstream neighbor | ATP11AUN |
C13orf35, SMABLO1 |
40.6kb | Anti-sense | Chr. 13 - 112647044 - 112684497 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map