Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
centrobin, centriole duplication and spindle assembly protein |
CNTROB |
LIP8, PP1221 |
-63.4kb | Anti-sense | Chr. 17 - 7932054 - 7949919 on NCBI Build GRCh38 |
| Downstream |
arachidonate 15-lipoxygenase, type B |
ALOX15B | 15-LOX-2 | 22.7kb | Anti-sense | Chr. 17 - 8039040 - 8049134 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map