Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich alpha-2-glycoprotein 1 | LRG1 |
HMFT1766, LRG |
-29.6kb | Sense | Chr. 19 - 4537215 - 4540024 on NCBI Build GRCh38 |
| Downstream | TNF alpha induced protein 8 like 1 | TNFAIP8L1 | TIPE1 | 67.5kb | Anti-sense | Chr. 19 - 4639505 - 4655568 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map