Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102723927 | LOC102723927 | -5.9kb | Sense | Chr. 2 - 241970683 - 241977276 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC728323 | LOC728323 | 23.2kb | Sense | Chr. 2 - 242088633 - 242160503 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map