Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serine/threonine kinase 32B | STK32B |
HSA250839, STK32, STKG6, YANK2 |
Sense | Chr. 4 - 5051323 - 5501001 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1587 | LINC01587 |
C4orf6, aC1 |
327.8kb | Sense | Chr. 4 - 5524569 - 5527801 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map