Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105369606 | LOC105369606 | -134.4kb | Anti-sense | Chr. 12 - 3300329 - 3327103 on NCBI Build GRCh38 | |
| Downstream | thyroid cancer-associated transcript 155 | THCAT155 | 22.2kb | Sense | Chr. 12 - 3483712 - 3492819 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map