Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | decapping mRNA 2 | DCP2 | NUDT20 | -377.7kb | Sense | Chr. 5 - 112976702 - 113022195 on NCBI Build GRCh38 |
| Downstream | testis specific serine kinase 1B | TSSK1B |
FKSG81, SPOGA4, STK22D, TSK1, TSSK1 |
3.8kb | Anti-sense | Chr. 5 - 113432554 - 113435031 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map