Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | decapping mRNA 2 | DCP2 | NUDT20 | -410.8kb | Sense | Chr. 5 - 112976702 - 113022195 on NCBI Build GRCh38 |
| Downstream | YTH domain containing 2 | YTHDC2 | CAHL | 77.3kb | Sense | Chr. 5 - 113513683 - 113595287 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map