Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ankyrin repeat domain 36B pseudogene 2 | ANKRD36BP2 | -1.6kb | Sense | Chr. 2 - 88765902 - 88806596 on NCBI Build GRCh38 | |
| Downstream | microRNA 4436a | MIR4436A | mir-4436a | 912.0 | Sense | Chr. 2 - 88812370 - 88812454 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map