| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
ENST00000521870.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
ENST00000521870.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
ENST00000521870.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500165524.hg.1 |
ENST00000521870.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500165524.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500165524.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500165524.hg.1 |
ENST00000521870.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500165524.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500165524.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500165524.hg.1 |
TC0500009361.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500165527.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500165527.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500165527.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500165527.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500165527.hg.1 |
TC0500009361.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500165526.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500165526.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500165526.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500165526.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500165526.hg.1 |
TC0500009361.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
ENST00000521870.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
ENST00000521870.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
ENST00000521870.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009361.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500165525.hg.1 |
ENST00000521870.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500165525.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500165525.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500165525.hg.1 |
ENST00000521870.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500165525.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500165525.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500165525.hg.1 |
TC0500009361.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500165522.hg.1 |
ENST00000521870.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500165522.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500165522.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500165522.hg.1 |
ENST00000521870.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500165522.hg.1 |
ENST00000522615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500165522.hg.1 |
NR_130737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500165522.hg.1 |
TC0500009361.hg.1 |
Clariom D TC |